missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 11338336
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338336 5 g 5g
11328336 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338336 Supplier Thermo Scientific Alfa Aesar Supplier No. L15584.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Reactant for Suzuki-Miyaura cross-couplings, NHC-Iron-catalyzed aerobic oxidative aromatic esterification of aldehydes, Palladium-catalyzed oxidative Heck-type reactions.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145240-28-4
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.038
MDL Number MFCD02093926
InChI Key UGZUUTHZEATQAM-UHFFFAOYSA-N
Synonym 4-butylphenyl boronic acid, 4-n-butylphenylboronic acid, 4-butylphenylboronicacid, 4-butylbenzeneboronic acid, 4-n-butylbenzeneboronic acid, 4-butylphenyl boranediol, boronic acid, 4-butylphenyl, pubchem18523, acmc-209cue, 4-butylphenylboranic acid
PubChem CID 4100860
IUPAC Name (4-butylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)CCCC)(O)O

Specifications

Melting Point 103°C to 106°C
Quantity 5 g
Beilstein 6920034
Solubility Information Soluble in methanol.
Formula Weight 178.04
Percent Purity 98%
Chemical Name or Material 4-n-Butylbenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.