missing translation for 'onlineSavingsMsg'
Learn More

4-n-Decylbiphenyl, 98%, Thermo Scientific™

Product Code. 11462678
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462678 5 g 5g
11452678 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462678 Supplier Thermo Scientific Alfa Aesar Supplier No. A16685.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
PDLC monomer. As chemical reagent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93972-02-2
Molecular Formula C22H30
Molecular Weight (g/mol) 294.482
MDL Number MFCD00102117
InChI Key YYBDFGOLQQVHDV-UHFFFAOYSA-N
Synonym 4-n-decylbiphenyl, 4-decyl-biphenyl, 4-decyl-1,1'-biphenyl, 4-decylbiphenyl, 4-decyl-1-phenylbenzene, maybridge1_002181, 1,1'-biphenyl, 4-decyl
PubChem CID 2801612
IUPAC Name 1-decyl-4-phenylbenzene
SMILES CCCCCCCCCCC1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 52°C to 54°C
Boiling Point 204°C to 210°C (1.5 mmHg)
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 294.48
Percent Purity 98%
Chemical Name or Material 4-n-Decylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.