missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Octyl-d17-phenol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-n-Octyl-d17-phenol |
| CAS | 1219794-55-4 |
| Molecular Formula | C14 D17 H5 O |
| Quantity | 0.05 g |
| Synonym | 4-Octylphenol D17 (octyl D17) |
| InChI Formula | InChI=1 S/C14H22O/c1-2-3-4-5-6-7-8-13-9-11-14(15)12-10-13/h9-12,15 H,2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])c1ccc(O)cc1 |
| IUPAC Name | 4-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecadeuteriooctyl)phenol |
| Molecular Weight (g/mol) | 223.43 |
| Formula Weight | 223.2738 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction