missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzaldehyde, 95%

Product Code. 11377286
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377286 1 g 1g
11387286 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377286 Supplier Thermo Scientific Alfa Aesar Supplier No. L09208.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6853-57-2 | C12H16O | 176.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6853-57-2
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00043599
InChI Key NQVZPRUSNWNSQH-UHFFFAOYSA-N
Synonym p-pentylbenzaldehyde, benzaldehyde, 4-pentyl, 4-n-pentylbenzaldehyde, p-amylbenzaldehyde, 4-n-amylbenzaldehyde, p-n-pentylbenzaldehyde, p-pentyl-benzaldehyde, p-n-pentylben-zaldehyde, p-pentylbenzaldehyde, i
PubChem CID 23290
IUPAC Name 4-pentylbenzaldehyde
SMILES CCCCCC1=CC=C(C=C1)C=O

Specifications

Boiling Point 142°C to 144°C (2 mmHg)
Refractive Index 1.5215
Quantity 1 g
Beilstein 1869642
Sensitivity Air Sensitive
Formula Weight 176.26
Percent Purity 95%
Chemical Name or Material 4-n-Pentylbenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.