missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenol, 98%

Product Code. 11448256
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448256 100 g 100g
11428256 5 g 5g
11438256 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448256 Supplier Thermo Scientific Alfa Aesar Supplier No. A15342.22
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 645-56-7 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-56-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002395
InChI Key KLSLBUSXWBJMEC-UHFFFAOYSA-N
Synonym 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
PubChem CID 12580
ChEBI CHEBI:34434
IUPAC Name 4-propylphenol
SMILES CCCC1=CC=C(C=C1)O

Specifications

Melting Point 20°C to 22°C
Density 0.983
Boiling Point 232°C
Flash Point 106°C (222°F)
Refractive Index 1.523
Quantity 100 g
UN Number UN3145
Beilstein 1363429
Formula Weight 136.19
Percent Purity 98%
Chemical Name or Material 4-n-Propylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.