missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2-(trifluoromethyl)benzonitrile, 98%

Product Code. 11372987
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372987 1 g 1g
11382987 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372987 Supplier Thermo Scientific Alfa Aesar Supplier No. L16318.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 320-47-8 | C8H3F3N2O2 | 216.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-47-8
Molecular Formula C8H3F3N2O2
Molecular Weight (g/mol) 216.119
MDL Number MFCD01075729
InChI Key AGKQJEFSEQHGTA-UHFFFAOYSA-N
Synonym 4-nitro-2-trifluoromethyl benzonitrile, 2-cyano-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitrobenzonitrile, benzonitrile, 4-nitro-2-trifluoromethyl, 4-nitro-2-trifluoromethyl benzenecarbonitrile, pubchem4821, acmc-1ctuy, buttpark 91\04-59, 4-cyano-3-trifluoromethyl nitrobenzene
PubChem CID 2775783
IUPAC Name 4-nitro-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)C#N

Specifications

Melting Point 50°C to 54°C
Quantity 1 g
UN Number UN3439
Beilstein 2281014
Formula Weight 216.12
Percent Purity 98%
Chemical Name or Material 4-Nitro-2-(trifluoromethyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.