missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)aniline, 98%

Código de producto. 11454874
Cambiar vista
Click to view available options
Quantity:
1 g
5 g
Tamaño de la unidad:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
11454874 5 g 5g
11444874 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11454874 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor A13798.06
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format
Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 393-11-3 | C7H5F3N2O2 | 206.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 393-11-3
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.12
MDL Number MFCD00014717
InChI Key UTKUVRNVYFTEHF-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride
PubChem CID 94955
IUPAC Name 4-nitro-3-(trifluoromethyl)aniline
SMILES NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O

Especificaciones

Melting Point 125°C to 128°C
Quantity 5 g
UN Number UN2811
Beilstein 2650702
Formula Weight 206.13
Percent Purity 98%
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)aniline
compliance-icons
Nombre del producto
  • 4-Nitro-3-(trifluoromethyl)aniline
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Nitro-3-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.