missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)phenol, 97%

Product Code. p-7044786
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398045 1 g 1g
11308055 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398045 Supplier Thermo Scientific Alfa Aesar Supplier No. L08228.03
Add to basket
This item is not returnable. View return policy

CAS: 88-30-2 | C7H4F3NO3 | 207.108 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-30-2
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.108
MDL Number MFCD00014714
InChI Key ZEFMBAFMCSYJOO-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl phenol, lamprecide, lamprecid, 5-hydroxy-2-nitrobenzotrifluoride, dowlap f, 3-trifluoromethyl-4-nitrophenol, phenol, 4-nitro-3-trifluoromethyl, usaf ma-6, caswell no. 890, tfm piscicide
PubChem CID 6931
IUPAC Name 4-nitro-3-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1O)C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 74°C to 77°C
Boiling Point 135°C to 138°C (0.01 mmHg)
Quantity 1 g
UN Number UN2811
Beilstein 2053698
Merck Index 14,9253
Formula Weight 207.11
Percent Purity 97%
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.