missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Chloroformate, TRC

Product Code. 30481516
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481516 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481516 Supplier Toronto Research Chemicals Supplier No. TRCN493645250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitrobenzyl Chloroformate
CAS 4457-32-3
Molecular Formula C8H6ClNO4
Quantity 250 g
Synonym Chloroformic Acid p-Nitrobenzyl Ester, 4-Nitrobenzyl Chloroformate, 4-Nitrobenzyloxycarbonyl Chloride, p-Nitrobenzyl Chloroformate, p-Nitrobenzyloxycarbonyl Chloride, p-Nitrocarbobenzoxy Chloride
InChI Formula InChI=1S/C8H6ClNO4/c9-8(11)14-5-6-1-3-7(4-2-6)10(12)13/h1-4H,5H2
SMILES [O-][N+](=O)c1ccc(COC(=O)Cl)cc1
IUPAC Name (4-nitrophenyl)methyl carbonochloridate
Molecular Weight (g/mol) 215.69
Formula Weight 214.9985
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.