Learn More
4-Nitrophenol, ≥99.5%, Solstice
Product Code. 15696940 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15696940 500 g Glass bottle 500g
15686940 25 g Glass bottle 25g
15676940 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15696940 Supplier Honeywell-Fluka Supplier No. 73560500G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O
Specifications
Melting Point 116°C
Boiling Point 279°C
Linear Formula O2NC6H4OH
UN Number UN1663
Beilstein 1281877
Solubility Information 14.8g/L at 25°C
Formula Weight 139.11g/mol
Percent Purity ≥99.5%
Packaging Glass bottle
Vapor Pressure 0.6mmHg (120°C)
Density 1.479g/cm3
Color Yellow
pH 4.4
Quantity 500 g
Physical Form Crystalline
Chemical Name or Material 4-Nitrophenol