missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenoxyacetic acid hydrazide, 98%, Thermo Scientific™

Product Code. 11492897
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492897 10 g 10g
11402907 50 g 50g
11344689 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492897 Supplier Thermo Scientific Alfa Aesar Supplier No. A15505.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75129-74-7
Molecular Formula C8H9N3O4
Molecular Weight (g/mol) 211.18
MDL Number MFCD00051973
InChI Key BDPKZWIKLVUGKS-UHFFFAOYSA-N
Synonym 2-4-nitrophenoxy acetohydrazide, 4-nitrophenoxyacetic acid hydrazide, acetic acid, 4-nitrophenoxy-, hydrazide, 2-4-nitrophenoxy ethanehydrazide, 4-nitrophenoxy acetic acid hydrazide, acetic acid,2-4-nitrophenoxy-, hydrazide, 2-4-nitro-phenoxy-acetic acid hydrazide
PubChem CID 156556
IUPAC Name 2-(4-nitrophenoxy)acetohydrazide
SMILES NNC(=O)COC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 185°C to 192°C
Quantity 10 g
Beilstein 2562952
Formula Weight 211.18
Percent Purity 98%
Chemical Name or Material 4-Nitrophenoxyacetic acid hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.