missing translation for 'onlineSavingsMsg'
Learn More

4-O-Desmethyl 4-O-Ethyl Trimethoprim, TRC

Product Code. 17948424
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17948424 25 mg 25mg
17929754 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17948424 Supplier Toronto Research Chemicals Supplier No. TRCD29570025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78025-68-0
Molecular Formula C15 H20 N4 O3
Molecular Weight (g/mol) 304.34
Synonym 5-(4-Ethoxy-3,5-dimethoxybenzyl)pyrimidine-2,4-diamine, Trimethoprim Imp. G (EP)
IUPAC Name 5-[(4-ethoxy-3,5-dimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES CCOc1c(OC)cc(Cc2cnc(N)nc2N)cc1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H20N4O3/c1-4-22-13-11(20-2)6-9(7-12(13)21-3)5-10-8-18-15(17)19-14(10)16/h6-8H,4-5H2,1-3H3,(H4,16,17,18,19)
Formula Weight 304.1535
Chemical Name or Material 5-(4-Ethoxy-3,5-dimethoxybenzyl)pyrimidine-2,4-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.