Learn More
4'-Octylacetophenone, TRC
Product Code. 30425559
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425559 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30425559 Supplier Toronto Research Chemicals Supplier No. TRCO268030100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4'-Octylacetophenone
CAS 10541-56-7
Molecular Formula C16H24O
Quantity 100 g
Synonym 1-(4-Octylphenyl)ethanone, NSC 168987, p-Octylacetophenone, p-n-Octylacetophenone
InChI Formula InChI=1S/C16H24O/c1-3-4-5-6-7-8-9-15-10-12-16(13-11-15)14(2)17/h10-13H,3-9H2,1-2H3
SMILES O=C(C)C1=CC=C(CCCCCCCC)C=C1
IUPAC Name 1-(4-octylphenyl)ethan-1-one
Molecular Weight (g/mol) 232.36
Formula Weight 232.18