missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-4-(p-tolyl)butanoic acid, 98%, BLD Pharm™

Product Code. 31005760
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31005760 100 g 100g
30998524 25 g 25g
31002599 5 g 5g
31004785 1 g 1g
30991567 500 g 500g
31003177 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31005760 Supplier BLD Pharm Supplier No. BD13132100g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4619-20-9
Molecular Formula C11H12O3
MDL Number MFCD00020541
Quantity 100 g
SMILES O=C(O)CCC(C1=CC=C(C)C=C1)=O
Molecular Weight (g/mol) 192.21
Percent Purity 98%
Chemical Name or Material 4-Oxo-4-(p-tolyl)butanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.