Learn More
4'-Phenoxyacetophenone, 98+%
Product Code. 11459553
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459553 25 g 25g
11469553 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11459553 Supplier Thermo Scientific Alfa Aesar Supplier No. A12392.14
 Restricted Item
In Stock
Add to basket
CAS: 5031-78-7 | C14H12O2 | 212.25 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5031-78-7
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00008744
InChI Key DJNIFZYQFLFGDT-UHFFFAOYSA-N
Synonym 4'-phenoxyacetophenone, 1-4-phenoxyphenyl ethanone, 4-phenoxyacetophenone, p-phenoxyacetophenone, 1-4-phenoxyphenyl ethan-1-one, ethanone, 1-4-phenoxyphenyl, 4-acetyldiphenyl ether, 1-4-phenoxyphenyl-1-ethanone, 1-4-phenoxyphenyl ethanon, pubchem13195
PubChem CID 236783
SMILES CC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Specifications
Melting Point 48°C to 52°C
Boiling Point 325°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2049295
IUPAC Name 1-(4-phenoxyphenyl)ethanone
Formula Weight 212.25
Percent Purity ≥98%
Chemical Name or Material 4'-Phenoxyacetophenone