missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-pyrrolidinone, TRC

Product Code. 30004656
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004656 250 mg 250mg
30003348 500 mg 500mg
30004425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004656 Supplier Toronto Research Chemicals Supplier No. TRCP336740250MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1198-97-6
Molecular Formula C10 H11 N O
Molecular Weight (g/mol) 161.2
Synonym 4-Phenyl-2-pyrrolidinone, 4-Phenyl-2-pyrrolidone, β-Phenyl-γ-aminobutyric Acid Lactam
IUPAC Name 4-phenylpyrrolidin-2-one
SMILES O=C1CC(CN1)c2ccccc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H11NO/c12-10-6-9(7-11-10)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,11,12)
Formula Weight 161.0841
Percent Purity >95%
Chemical Name or Material 4-Phenyl-2-pyrrolidinone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.