missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pregnen-17alpha,21-diol-3,11,20-trione-1,2-d2, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Cortisone-1,2-D2 |
| CAS | 2687960-86-5 |
| Molecular Formula | C21 D2 H26 O5 |
| Quantity | 0.005 g |
| Synonym | Pregn-4-ene-3,11,20-trione-1,2-d2, 17,21-dihydroxy- (ACI), 17,21-Dihydroxypregn-4-ene-3,11,20-trione-1,2-d2 (ACI), Cortisone-1,2-D2, Cortisone-d2, 17-Hydroxy-11-dehydrocorticosterone-1,2-d2 |
| InChI Formula | InChI=1 S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-15,18,22,26 H,3-8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1/i5D,7 D/t5?,7?,14-,15-,18+,19-,20-,21- |
| SMILES | [2 H]C1C([2 H])[C@]2(C)[C@@H]3 [C@@H](CCC2=CC1=O)[C@@H]4 CC[C@](O)(C(=O)CO)[C@@]4(C)CC3=O |
| IUPAC Name | (8 S,9 S,10 R,13 S,14 S,17 R)-1,2-dideuterio-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione |
| Molecular Weight (g/mol) | 362.4563 |
| Formula Weight | 362.2062 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction