missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-3-thiosemicarbazide, 97%

Product Code. 11382946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382946 5 g 5g
11372946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382946 Supplier Thermo Scientific Alfa Aesar Supplier No. L10704.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 13431-39-5 | C5H13N3S | 147.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of antiproliferative activity of N-substituted 2-amino-5-(2, 4-dihydroxyphenyl)-1, 3, 4-thiadiazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13431-39-5
Molecular Formula C5H13N3S
Molecular Weight (g/mol) 147.24
MDL Number MFCD00041304
InChI Key ZUWRCNZOBNETMU-UHFFFAOYSA-N
Synonym n-tert-butyl hydrazinecarbothioamide, 4-tert-butyl-3-thiosemicarbazide, 3-amino-1-tert-butylthiourea, n-tert-butylhydrazinecarbothioamide, 4-tert-butylthiosemicarbazide, hydrazinecarbothioamide,n-1,1-dimethylethyl, tert-butyl amino hydrazinomethane-1-thione, acmc-20a36m, n1-tert-butyl hydrazine-1-carbothioamide, 4-1,1-dimethylethyl-3-thiosemicarbazide
PubChem CID 737275
IUPAC Name 1-amino-3-tert-butylthiourea
SMILES CC(C)(C)NC(=S)NN

Specifications

Melting Point 139°C to 140°C
Quantity 5 g
Beilstein 507219
Solubility Information Insoluble in water.
Formula Weight 147.24
Percent Purity 97%
Chemical Name or Material 4-tert-Butyl-3-thiosemicarbazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.