missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzeneboronic acid, 97%

Product Code. 11485857
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485857 1 g 1g
11495857 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485857 Supplier Thermo Scientific Alfa Aesar Supplier No. B24408.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 123324-71-0 | C10H15BO2 | 178.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123324-71-0
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.04
MDL Number MFCD01009697
InChI Key MNJYZNVROSZZQC-UHFFFAOYSA-N
Synonym 4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid, 4-tert-butyl-phenyl boronic acid, 4-tert-butylphenyl boranediol, p-t-butylphenylboronic acid, 4-tertbutylphenylboronic acid
PubChem CID 2734320
IUPAC Name (4-tert-butylphenyl)boronic acid
SMILES CC(C)(C)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 191°C to 196°C
Quantity 1 g
Beilstein 5330959
Formula Weight 178.04
Percent Purity 97%
Chemical Name or Material 4-tert-Butylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.