missing translation for 'onlineSavingsMsg'
Learn More

4-Thien-3-ylbenzonitrile, ≥97%, Thermo Scientific™

Product Code. 10412844
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10412844 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10412844 Supplier Thermo Scientific Maybridge Supplier No. CC48716DA
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 172507-33-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H7NS
MDL Number MFCD00115000
Quantity 1 g
Synonym 4-3-thienyl benzonitrile, 4-thiophen-3-yl benzonitrile, 4-thien-3-ylbenzonitrile, benzonitrile, 4-3-thienyl, 4-3-thienyl benzenecarbonitrile
InChI Key GRAMVGQZHXAXHP-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C#N)C2=CSC=C2
IUPAC Name 4-thiophen-3-ylbenzonitrile
Molecular Weight (g/mol) 185.244
PubChem CID 4130333
Formula Weight 185.25
Percent Purity 97%
Chemical Name or Material 4-Thien-3-ylbenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.