missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl isocyanate, 97%

Product Code. 11349449
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349449 25 g 25g
11440501 1 g 1g
11450501 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349449 Supplier Thermo Scientific Alfa Aesar Supplier No. B20033.14
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35037-73-1 | C8H4F3NO2 | 203.12 g/mol

4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine

Solubility
Reacts with water.

Notes
Moisture Sensitive. Protect from heat. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas. Incompatible with alcohols, amines and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35037-73-1
Molecular Formula C8H4F3NO2
Molecular Weight (g/mol) 203.12
MDL Number MFCD00035702
InChI Key LGPKFIGMLPDYEA-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
PubChem CID 92298
IUPAC Name 1-isocyanato-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1N=C=O)OC(F)(F)F

Specifications

Melting Point -29°C
Density 1.346
Boiling Point 78°C (22 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.458
Quantity 25 g
UN Number UN2206
Beilstein 977669
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 203.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.