missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethoxybenzyl Bromide, TRC

Product Code. 30450835
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450835 Supplier Toronto Research Chemicals Supplier No. TRCT7744401G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Trifluoromethoxybenzyl Bromide
CAS 50824-05-0
Molecular Formula C8H6BrF3O
Quantity 1 g
Synonym 1-Bromomethyl-4-trifluoromethoxybenzene, 4-(Bromomethyl)phenyl Trifluoromethyl Ether, 4-(Trifluoromethoxy)-1-(bromomethyl)-benzene, 4-Trifluoromethoxybenzyl Bromide, p-Trifluoromethoxybenzyl Bromide
InChI Formula InChI=1S/C8H6BrF3O/c9-5-6-1-3-7(4-2-6)13-8(10,11)12/h1-4H,5H2
SMILES FC(F)(F)OC1=CC=C(CBr)C=C1
IUPAC Name 1-(bromomethyl)-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol) 255.03
Formula Weight 253.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.