missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)aniline Hydrochloride, Mikromol™

Product Code. 30052135
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052135 25 mg 25mg
30052084 100 mg 100mg
30461246 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052135 Supplier Mikromol Supplier No. MM0859.010025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(Trifluoromethyl)aniline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Trifluoromethyl)aniline Hydrochloride
CAS 90774-69-9
Analyte or Component Names 4-(Trifluoromethyl)aniline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C7 H6 F3 N . Cl H
Certifications/Compliance ISO 17025
Synonym Leflunomide Imp. A (EP), Leflunomide USP Related Compound A, Leflunomide USP RC A, 4-(Trifluoromethyl)aniline Hydrochloride, Leflunomide Imp. A (EP) as Hydrochloride, Leflunomide Related Compound A as Hydrochloride, Leflunomide Impurity A as Hydrochloride
InChI Formula InChI=1S/C7H6F3N.ClH/c8-7(9,10)5-1-3-6(11)4-2-5;/h1-4H,11H2;1H
SMILES Cl.Nc1ccc(cc1)C(F)(F)F
IUPAC Name 4-(trifluoromethyl)aniline;hydrochloride
Molecular Weight (g/mol) 197.59
Quantity 25 mg
Formula Weight 197.0219 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.