missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzenesulfonyl chloride, 98%

Product Code. 11479713
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479713 1 g 1g
11489713 5 g 5g
11499713 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479713 Supplier Thermo Scientific Alfa Aesar Supplier No. A12907.03
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2991-42-6 | C7H4ClF3O2S | 244.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2991-42-6
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.61
MDL Number MFCD00042422
InChI Key OZDCZHDOIBUGAJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzene-1-sulfonyl chloride, 4-trifluoromethyl benzene sulfonyl chloride, p-trifluoromethylbenzenesulfonyl chloride, 4-trifluoromethyl benzenesulfonylchloride, 4-trifluoromethyl-benzenesulfonyl chloride, 4-trifluoromethyl phenylsulfonyl chloride, p-trifluoromethyl benzenesulfonyl chloride, 4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethyl
PubChem CID 2777399
IUPAC Name 4-(trifluoromethyl)benzenesulfonyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 28°C to 34°C
Boiling Point 118°C to 120°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Beilstein 2113604
Formula Weight 244.62
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.