missing translation for 'onlineSavingsMsg'
Learn More

4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 15499265
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
870238-65-6
C17H25BO2Si
300.28
MFCD08706286
CIKKODWPGJIJQG-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-4-trimethylsilanylethynyl-phenyl-1,3,2 dioxaborolane, 4-trimethylsilyl ethynyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-4-2-trimethylsilyl ethynyl phenyl, trimethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, 4,4,5,5-tetramethyl-2-4-trimethylsilyl ethynyl phenyl-1,3,2-dioxaborolane, amtb200, 4-trimethylsilyl ethynyl benzeneboronic acid pinacol ester, 2-4-trimethylsilylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
17999152
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#C[Si](C)(C)C
This item is not returnable. View return policy
870238-65-6
C17H25BO2Si
300.28
MFCD08706286
CIKKODWPGJIJQG-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-4-trimethylsilanylethynyl-phenyl-1,3,2 dioxaborolane, 4-trimethylsilyl ethynyl phenylboronic acid pinacol ester, 1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-4-2-trimethylsilyl ethynyl phenyl, trimethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl silane, 4,4,5,5-tetramethyl-2-4-trimethylsilyl ethynyl phenyl-1,3,2-dioxaborolane, amtb200, 4-trimethylsilyl ethynyl benzeneboronic acid pinacol ester, 2-4-trimethylsilylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
17999152
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#C[Si](C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
IUPAC Name trimethyl({2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl})silane
Formula Weight 300.28
Percent Purity 97%
Chemical Name or Material 4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'