Learn More
Cellpath FiloSlide Plastic Slide Wallets
Product Code. 30226296
Change view
Click to view available options
Includes:
6 microscope slide pockets, 1 document pocket (220 x 150mm), 1 index pocket; Index strips
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
30226296 6 microscope slide pockets, 1 document pocket (220 x 150mm), 1 index pocket; Index strips Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30226296 Supplier Cellpath Supplier No. WEP060301A
  • ONLINE EXCLUSIVE PRICE
    24.92€
    17.98€ / Pack of 10
    Save 6.94€
    27% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for building reference/teaching sets of slides
  • Superior slide protection
  • Document wallet on reverse to store paperwork
  • Easy to use
  • FiloSlide 4, 5, 6 Wallets made from clear flexible plastic
FiloSlide 4 slide wallet:
  • Four microscope slide pockets, one document pocket 103 x 153mm (4 x 6 in.)
  • Combination pack of 100 wallets and one FiloSlide cabinet also available
FiloSlide 5 slide wallet:
  • Five microscope slide pockets, one document pocket
  • Also available in combination pack of 100 wallets and one FiloSlide cabin
FiloSlide 6 Slide wallet:
  • Six microscope slide pockets and one document pocket 220 x 150mm (8.6 x 6 in.) and one index pocket
  • Each pack of 10 FiloSlide 6 wallets is supplied with 50 color coded index cards (10 each of red, blue, green, white, yellow)
  • Combination pack of 150 wallets and one FiloSlide cabinet is available
Specifications
Slide Capacity 6
Includes 6 microscope slide pockets, 1 document pocket (220 x 150mm), 1 index pocket; Index strips
Material Plastic