missing translation for 'onlineSavingsMsg'
Learn More

(4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic Acid (Mycophenolic Acid), Mikromol™

Product Code. 18062110
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18062110 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18062110 Supplier Mikromol Supplier No. MM1001.060250
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic Acid (Mycophenolic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mycophenolic acid
CAS 24280-93-1
Analyte or Component Names Mycophenolic acid
Shipping Condition Room Temperature
Molecular Formula C17 H20 O6
Certifications/Compliance ISO 17034
Synonym (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic Acid, Mycophenolate Mofetil Imp. F (EP), Mycophenolic Acid, Mycophenolate Mofetil Impurity F
InChI Formula InChI=1S/C17H20O6/c1-9(5-7-13(18)19)4-6-11-15(20)14-12(8-23-17(14)21)10(2)16(11)22-3/h4,20H,5-8H2,1-3H3,(H,18,19)/b9-4+
SMILES COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)\CCC(=O)O
IUPAC Name (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
Molecular Weight (g/mol) 320.34
Quantity 250 mg
Formula Weight 320.126 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.