missing translation for 'onlineSavingsMsg'
Learn More

4H-Thieno[3,2-b]pyrrole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15474528
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474528 5 g 5g
15464528 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474528 Supplier Thermo Scientific Alfa Aesar Supplier No. H58200.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39793-31-2 | Molecular Formula: C7H5NO2S | Molecular Weight: 167.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39793-31-2
Molecular Formula C7H5NO2S
Molecular Weight (g/mol) 167.182
MDL Number MFCD07368542
InChI Key PMHDSACGRKBACK-UHFFFAOYSA-N
Synonym 4h-thieno 3,2-b pyrrole-5-carboxylic acid, 4h-thieno 3,2-b pyrole-5-carboxylic acid, 4wl, d-amino acid oxidase inhibitor, 4h-thieno 3,2-b pyrrole-5-carboxylicacid, thiopheno 3,2-b pyrrole-5-carboxylic acid, 4h-thieno 2,3-d pyrrole-5-carboxylic acid, 4h-thieno 3 pound not2-b pyrrole-5-carboxylic acid
PubChem CID 4961254
IUPAC Name 4H-thieno[3,2-b]pyrrole-5-carboxylic acid
SMILES C1=CSC2=C1NC(=C2)C(=O)O

Specifications

Melting Point 180°C
Quantity 5 g
Formula Weight 167.19
Percent Purity 97%
Chemical Name or Material 4H-Thieno[3,2-b]pyrrole-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.