missing translation for 'onlineSavingsMsg'
Learn More

(4R)-5-(3',4'-Dihydroxyphenyl)-Gamma-valerolactone ((-)-Epicatechin Metabolite), TRC

Product Code. 30003743
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003743 2.5 mg 2.50mg
30008689 5 mg 5mg
30007463 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003743 Supplier Toronto Research Chemicals Supplier No. TRCD4545252.5MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191666-22-5
Molecular Formula C11 H12 O4
Molecular Weight (g/mol) 208.21
Synonym (4R)-5-(3',4'-Dihydroxyphenyl)-valerolactone, (-)-Epicatechin metabolite
IUPAC Name (5R)-5-[(3,4-dihydroxyphenyl)methyl]oxolan-2-one
SMILES Oc1ccc(C[C@H]2CCC(=O)O2)cc1O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C11H12O4/c12-9-3-1-7(6-10(9)13)5-8-2-4-11(14)15-8/h1,3,6,8,12-13H,2,4-5H2/t8-/m1/s1
Formula Weight 208.0736
Chemical Name or Material (4R)-5-(3',4'-Dihydroxyphenyl)-valerolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.