missing translation for 'onlineSavingsMsg'
Learn More

5-[1-(2,3-Dimethylphenyl)ethenyl]-1H-imidazole, TRC

Product Code. 30138167
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138167 500 mg 500mg
30134762 1 g 1g
30011241 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138167 Supplier Toronto Research Chemicals Supplier No. TRCD476965500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1021949-47-2
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.26
Synonym Vinyl Medetomidine
SMILES CC1=C(C)C(=CC=C1)C(=C)C2=CN=CN2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H14N2/c1-9-5-4-6-12(10(9)2)11(3)13-7-14-8-15-13/h4-8H,3H2,1-2H3,(H,14,15)
Formula Weight 198.1157
Chemical Name or Material 5-[1-(2,3-Dimethylphenyl)ethenyl]-1H-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.