missing translation for 'onlineSavingsMsg'
Learn More

5-(1,2-Dithiolan-3-yl)pentanoic acid, 97%, BLD Pharm™

Product Code. 31033261
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Unit Size:
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31033261 100 g -
31018548 25 g -
31026843 1 kg -
31021525 5 g 5g
31026231 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31033261 Supplier BLD Pharm Supplier No. BD126427100g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1077-28-7
Molecular Formula C8H14O2S2
MDL Number MFCD00005474
Quantity 100 g
SMILES C(C1CCSS1)CCCC(O)=O
Molecular Weight (g/mol) 206.33
Percent Purity 97%
Chemical Name or Material 5-(1,2-Dithiolan-3-yl)pentanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.