missing translation for 'onlineSavingsMsg'
Learn More

5,11-Dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one, TRC

Product Code. 30381517
Change view
Click to view available options
Quantity:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381517 2.5 g
30384759 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381517 Supplier Toronto Research Chemicals Supplier No. TRCD4467202.5G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885-70-1
Molecular Formula C12H9N3O
Molecular Weight (g/mol) 211.22
SMILES O=C1Nc2cccnc2Nc3ccccc13

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H9N3O/c16-12-8-4-1-2-5-9(8)14-11-10(15-12)6-3-7-13-11/h1-7H,(H,13,14)(H,15,16)
Formula Weight 211.0746
Chemical Name or Material 5,11-Dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.