missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Fluorophenyl)-N-methyl-1H-pyrrole-3-methanamine, TRC

Product Code. 30135742
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135742 500 mg 500mg
17991775 1 g 1g
17924975 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135742 Supplier Toronto Research Chemicals Supplier No. TRCD632263500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1610043-62-3
Molecular Formula C13 H10 F3 N O
Molecular Weight (g/mol) 253.22
Synonym 1-(5-(2-Fluorophenyl)-1H-pyrrol-3-yl)-N-methylmethanamine
IUPAC Name 4-phenoxy-2-(trifluoromethyl)aniline
SMILES Nc1ccc(Oc2ccccc2)cc1C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H10F3NO/c14-13(15,16)11-8-10(6-7-12(11)17)18-9-4-2-1-3-5-9/h1-8H,17H2
Formula Weight 253.071
Percent Purity >95%
Chemical Name or Material 5-(2-Fluorophenyl)-N-methyl-1H-pyrrole-3-methanamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.