missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Pyridinylsulfanyl)-2-furaldehyde, 95+%, Thermo Scientific™

Product Code. 10071524
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10071524 5 g 5g
10432043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071524 Supplier Thermo Scientific Maybridge Supplier No. MO07099DE
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 709635-68-7
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.231
InChI Key NWJZBQDKKLHSBP-UHFFFAOYSA-N
Synonym 5-2-pyridinylsulfanyl-2-furaldehyde, 5-pyridin-2-ylthio furan-2-carbaldehyde, 5-pyridin-2-ylsulfanyl furan-2-carbaldehyde, 5-pyridin-2-ylsulfanyl-furan-2-carbaldehyde, 5-pyridin-2-ylthio-2-furaldehyde, 5-2-pyridylthio furan-2-carbaldehyde, 5-2-pyridylsulfanyl furan-2-carbaldehyde, 2-furancarboxaldehyde,5-2-pyridinylthio, 2-furancarboxaldehyde, 5-2-pyridinylthio
PubChem CID 2794784
IUPAC Name 5-pyridin-2-ylsulfanylfuran-2-carbaldehyde
SMILES C1=CC=NC(=C1)SC2=CC=C(O2)C=O

Specifications

Assay Percent Range 95%
Quantity 5 g
Formula Weight 205.24
Percent Purity 95%
Chemical Name or Material 5-(2-Pyridinylsulfanyl)-2-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.