missing translation for 'onlineSavingsMsg'
Learn More

5-(3,4-dihydroxyphenyl)pentanoic Acid, TRC

Product Code. 30135401
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135401 25 mg 25mg
30010640 5 mg 5mg
30011488 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135401 Supplier Toronto Research Chemicals Supplier No. TRCD45850525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31129-94-9
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
SMILES OC(=O)CCCCc1ccc(O)c(O)c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H14O4/c12-9-6-5-8(7-10(9)13)3-1-2-4-11(14)15/h5-7,12-13H,1-4H2,(H,14,15)
Formula Weight 210.0892
Chemical Name or Material 5-(3,4-dihydroxyphenyl)pentanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.