missing translation for 'onlineSavingsMsg'
Learn More

5,3',4'-Trihydroxyflavone (>85%), TRC

Product Code. 30457111
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
Unit Size:
200mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457111 25 mg 25mg
30450024 50 mg 50mg
30453631 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457111 Supplier Toronto Research Chemicals Supplier No. TRCT89668525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19852-25-6
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
IUPAC Name 2-(3,4-dihydroxyphenyl)-5-hydroxychromen-4-one
SMILES Oc1ccc(cc1O)C2=CC(=O)c3c(O)cccc3O2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H10O5/c16-9-5-4-8(6-11(9)18)14-7-12(19)15-10(17)2-1-3-13(15)20-14/h1-7,16-18H
Formula Weight 270.0528
Chemical Name or Material 5,3',4'-Trihydroxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.