missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Methyl-1,3-dioxolan-2-yl)-1,3-benzodioxole, TRC

Product Code. 30135957
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135957 1 g 1g
30138064 500 mg 500mg
30012829 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135957 Supplier Toronto Research Chemicals Supplier No. TRCM4510031G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61683-99-6
Molecular Formula C11 H12 O4
Molecular Weight (g/mol) 208.211
Synonym 1,3-Benzodioxole, 5-(4-methyl-1,3-dioxolan-2-yl)- (9CI, ACI), 1,3-Dioxolane, 4-methyl-2-(3,4-methylenedioxyphenyl)- (6CI), 5-(4-Methyl-1,3-dioxolan-2-yl)-1,3-benzodioxole (ACI), NSC 406081, Piperonal propylene glycol acetal
IUPAC Name 5-(4-methyl-1,3-dioxolan-2-yl)-1,3-benzodioxole
SMILES CC1COC(O1)c2ccc3OCOc3c2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H12O4/c1-7-5-12-11(15-7)8-2-3-9-10(4-8)14-6-13-9/h2-4,7,11H,5-6H2,1H3
Formula Weight 208.074
Chemical Name or Material 5-(4-Methyl-1,3-dioxolan-2-yl)-1,3-benzodioxole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.