Learn More
5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid |
| CAS | 4242-18-6 |
| Molecular Formula | C11 H12 O2 |
| Quantity | 2 g |
| Synonym | 1-Naphthalenecarboxylic acid','5,6,7,8-tetrahydro-, 1-Naphthoic acid','5,6,7,8-tetrahydro- (6CI,7CI,8CI), 5,6,7,8-Tetrahydro-1-naphthalenecarboxylic acid, 5,6,7,8-Tetrahydro-1-naphthoic acid, 5,6,7,8-Tetrahydronaphthalen-1-carboxylic acid, NSC 44874, 5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid |
| InChI Formula | InChI=1S/C11H12O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h3,5,7H,1-2,4,6H2,(H,12,13) |
| SMILES | OC(=O)c1cccc2CCCCc12 |
| IUPAC Name | 5,6,7,8-tetrahydronaphthalene-1-carboxylic acid |
| Molecular Weight (g/mol) | 176.21 |
| Formula Weight | 176.0837 |