missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chlorobenzoic acid, 97%, Thermo Scientific™

Product Code. 15462142 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462142 100 g 100g
15452142 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462142 Supplier Thermo Scientific Acros Supplier No. 335251000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

May develop some turbidity or precipitate may partially crystallize

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-54-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007845
InChI Key GVCFFVPEOLCYNN-UHFFFAOYSA-N
Synonym 2-chloro-5-aminobenzoic acid, benzoic acid, 5-amino-2-chloro, 3-amino-6-chlorobenzoic acid, 5-amino-2-chloro-benzoic acid, 5-amino-2-chlorobenzoicacid, 3-carboxy-4-chloroaniline, 5-amino-2-chloro benzoic acid, zlchem 462, pubchem4654, acmc-209r1l
PubChem CID 37879
IUPAC Name 5-amino-2-chlorobenzoic acid
SMILES NC1=CC=C(Cl)C(=C1)C(O)=O

Specifications

Melting Point 184.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 100 g
Formula Weight 171.58
Percent Purity 97%
Chemical Name or Material 5-Amino-2-chlorobenzoic acid
compliance-icons
Product Identifier
  • 5-Amino-2-chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.