missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-N,N’-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide, TRC

Product Code. 30138205
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138205 50 mg 50mg
30136906 250 mg 250mg
30007484 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138205 Supplier Toronto Research Chemicals Supplier No. TRCA60192050MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76801-93-9
Molecular Formula C14 H18 I3 N3 O6
Molecular Weight (g/mol) 705.02
Synonym 3,5-Bis(2,3-dihydroxypropylaminocarbonyl)-2,4,6-triiodoaniline, 5-Amino-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide, 5-Amino-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide, Iohexol Related Compound B, Ioversol Related Compound A
IUPAC Name 5-amino-1-N,3-N-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide
SMILES Nc1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H18I3N3O6/c15-9-7(13(25)19-1-5(23)3-21)10(16)12(18)11(17)8(9)14(26)20-2-6(24)4-22/h5-6,21-24H,1-4,18H2,(H,19,25)(H,20,26)
Formula Weight 704.833
Chemical Name or Material 5-Amino-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.