missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindole, 97%

Product Code. 11405837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405837 1 g 1g
11415837 5 g 5g
11425837 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405837 Supplier Thermo Scientific Alfa Aesar Supplier No. B24391.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5192-03-0 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5192-03-0
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00005679
InChI Key ZCBIFHNDZBSCEP-UHFFFAOYSA-N
Synonym 5-aminoindole, indol-5-ylamine, indole, 5-amino, unii-q732pg0y16, 1h-indol-5-ylamine, indole-5-ylamine, 5-indolamine, 5-amino indole, 5-amino-indole
PubChem CID 78867
ChEBI CHEBI:33067
IUPAC Name 1H-indol-5-amine
SMILES NC1=CC=C2NC=CC2=C1

Specifications

Melting Point 130°C to 134°C
Quantity 1 g
Beilstein 112348
Sensitivity Air sensitive
Formula Weight 132.17
Percent Purity 97%
Chemical Name or Material 5-Aminoindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.