missing translation for 'onlineSavingsMsg'
Learn More

5-Aminonicotinic acid, 95%, Thermo Scientific™

Product Code. 10093383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10093383 5 g Glass bottle 5g
10562992 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10093383 Supplier Thermo Scientific Acros Supplier No. 361140050
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 24242-19-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00129116
InChI Key BYIORJAACCWFPU-UHFFFAOYSA-N
Synonym 5-aminonicotinic acid, 5-amino-nicotinic acid, 5-aminonicotinicacid, 3-pyridinecarboxylic acid, 5-amino, 5-amino nicotinic acid, 5-amino-3-carboxypyridine, 5-amino-3-pyridinecarboxylic acid, zlchem 307, pubchem15914, acmc-209ga2
PubChem CID 354316
ChEBI CHEBI:68578
IUPAC Name 5-aminopyridine-3-carboxylic acid
SMILES C1=C(C=NC=C1N)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 138.13
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Aminonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.