missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dimethoxypyrimidine, 97%, Thermo Scientific™

Product Code. 11956561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11956561 1 g 1g
11966561 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11956561 Supplier Thermo Scientific Acros Supplier No. 443390010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 56686-16-9
InChI Key QEZIMQMMEGPYTR-UHFFFAOYSA-N
Synonym 2,4-dimethoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxy-1,3-diazine, 5-bromo-2,4-bis methyloxy pyrimidine, 5-bromo-2,4-dimethoxy-pyrimidine, pubchem5281, acmc-209lu1, ksc495s0l, 5-bromo-2,4-dimethoxypyrimidine, 5-bromo-2,4-dimethoxypyrimidine;
PubChem CID 255719
IUPAC Name 5-bromo-2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1Br)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 62°C to 67°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H7BrN2O2
Quantity 1 g
Molecular Weight (g/mol) 219.038
Formula Weight 219.04
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 5-Bromo-2, 4-dimethoxypyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2,4-dimethoxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.