missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloroanisole, 97%, Thermo Scientific™

Product Code. 15491828
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491828 250 mg 250mg
15471828 1 g 1g
15481828 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491828 Supplier Thermo Scientific Alfa Aesar Supplier No. H55533.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-chloroanisole is used to produce 4-chloro-3-methoxy-benzonitrile at the temperature of 110°C. It will need solvent dimethylformamide

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16817-43-9
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00078659
InChI Key UAMVKOTWSHJOSY-UHFFFAOYSA-N
Synonym 5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq
PubChem CID 2769567
IUPAC Name 4-bromo-1-chloro-2-methoxybenzene
SMILES COC1=C(Cl)C=CC(Br)=C1

Specifications

Melting Point 27°C to 28°C
Density 1.631
Boiling Point 240°C to 245°C
Flash Point 113°C (235°F)
Refractive Index 1.587
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 221.48
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-chloroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.