missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzylamine hydrochloride, 97%

Product Code. 11470628
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470628 25 g 25g
15463277 1 g 1g
11460628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470628 Supplier Thermo Scientific Alfa Aesar Supplier No. B25176.14
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 202865-69-8 | C7H8BrFN | 205.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-69-8
Molecular Formula C7H8BrFN
Molecular Weight (g/mol) 205.05
MDL Number MFCD00143426
InChI Key PDKBJZGGXHTHNC-UHFFFAOYSA-O
Synonym 5-bromo-2-fluorobenzylamine hydrochloride, 5-bromo-2-fluorophenyl methanamine hydrochloride, 5-bromo-2-fluorobenzylamine hcl, 5-bromo-2-fluorophenyl methanamine hcl, 1-5-bromo-2-fluorophenyl methanamine hydrochloride, pubchem3227, acmc-1cpb5, rarechem al bd 0325, akos bbv-005136, 5-bromo-2-fluorobenzyl amine hcl
PubChem CID 2724903
SMILES [NH3+]CC1=CC(Br)=CC=C1F

Specifications

Melting Point 221°C to 226°C
Quantity 25 g
Sensitivity Hygroscopic
IUPAC Name (5-bromo-2-fluorophenyl)methanamine;hydrochloride
Formula Weight 240.5
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-fluorobenzylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.