missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, Maybridge

Product Code. 10802231
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10802231 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10802231 Supplier Thermo Scientific Maybridge Supplier No. MAY00186DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 749875-16-9
Packaging Amber Glass Bottle
Molecular Formula C7H3BrF3NO2
MDL Number MFCD10687236
Quantity 1 g
Synonym 5-bromo-2-trifluoromethyl isonicotinic acid, 5-bromo-2-trifluoromethyl pyridine-4-carboxylic acid, 5-bromo-2-trifluoromethyl-isonicotinic acid, 4-pyridinecarboxylic acid, 5-bromo-2-trifluoromethyl, 5-bromo-2-trifluoromethyl isonicotinicacid, 3-bromo-6-trifluoromethyl pyridine-4-carboxylic acid
InChI Key XNVAVQMVAYUTBA-UHFFFAOYSA-N
SMILES OC(=O)C1=CC(=NC=C1Br)C(F)(F)F
IUPAC Name 5-bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Molecular Weight (g/mol) 270.01
PubChem CID 28306936
Formula Weight 270.01
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.