missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoisoquinoline, 97%, ACROS Organics™

Product Code. 10544964 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10544964 5 g Glass bottle 5g
10336414 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10544964 Supplier Thermo Scientific Acros Supplier No. 372750050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 34784-04-8
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD01646405
InChI Key CYJZJGYYTFQQBY-UHFFFAOYSA-N
Synonym isoquinoline, 5-bromo, 5-bromo-isoquinoline, 5-bromo isoquinoline, 5-bromoisoquinoline?, pubchem6214, acmc-1adiy, 5-bromoisoquinoline, ksc226k2l, buttpark 44\07-76, 5-bromoisoquinoline 250mg
PubChem CID 736487
IUPAC Name 5-bromoisoquinoline
SMILES BrC1=C2C=CN=CC2=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 208.06
Chemical Name or Material 5-Bromoisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.