missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoisoquinoline, 98%

Product Code. 11336419
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336419 5 g 5g
11325425 250 mg 250mg
11315425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336419 Supplier Thermo Scientific Alfa Aesar Supplier No. L19632.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 34784-04-8 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34784-04-8
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD01646405
InChI Key CYJZJGYYTFQQBY-UHFFFAOYSA-N
Synonym isoquinoline, 5-bromo, 5-bromo-isoquinoline, 5-bromo isoquinoline, 5-bromoisoquinoline?, pubchem6214, acmc-1adiy, 5-bromoisoquinoline, ksc226k2l, buttpark 44\07-76, 5-bromoisoquinoline 250mg
PubChem CID 736487
IUPAC Name 5-bromoisoquinoline
SMILES BrC1=C2C=CN=CC2=CC=C1

Specifications

Melting Point 81°C to 83°C
Boiling Point 95°C to 97°C (0.1 mmHg)
Quantity 5 g
Beilstein 114468
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 5-Bromoisoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.