missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromosalicylic acid, 98%, Thermo Scientific™

Product Code. 11469866
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469866 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11469866 Supplier Alfa Aesar™ Supplier No. B20338.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromosalicylic acid is used as organic synthesis. The cross-coupling reaction of 5-bromosalicylic acid with alkyl and aryl Grignard reagents provides a navel efficient synthesis of substituted salicylic acids.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-55-4
Molecular Formula C7H5BrO3
Molecular Weight (g/mol) 217.02
MDL Number MFCD00002455
InChI Key IEJOONSLOGAXNO-UHFFFAOYSA-N
Synonym 5-bromosalicylic acid, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo, 2-hydroxy-5-bromobenzoic acid, benzoic acid, 3-bromo-6-hydroxy, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromsalicylic acid, 5-bromosalicyclic acid, acmc-209r1n
PubChem CID 6972
IUPAC Name 5-bromo-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(Br)=CC=C1O

Specifications

Melting Point 166°C to 170°C
Quantity 50 g
Beilstein 2209121
Formula Weight 217.02
Percent Purity 98%
Chemical Name or Material 5-Bromosalicylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.