missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methyl-4-nitro-1H-imidazole, Mikromol™

Product Code. 30460645
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460645 4 x 25 mg -
17901016 25 mg 25mg
17934046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30460645 Supplier Mikromol Supplier No. MM1532.0100254AMP
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Chloro-1-methyl-4-nitro-1H-imidazole, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-1-Methyl-4-Nitroimidazole
CAS 4897-25-0
Analyte or Component Names 5-Chloro-1-Methyl-4-Nitroimidazole
Shipping Condition Room Temperature
Molecular Formula C4 H4 Cl N3 O2
Certifications/Compliance ISO 17025
Synonym Azathioprine Imp. C (EP), 5-Chloro-1-methyl-4-nitroimidazole, Azathioprine Impurity C
InChI Formula InChI=1S/C4H4ClN3O2/c1-7-2-6-4(3(7)5)8(9)10/h2H,1H3
Solution Type Neat
SMILES Cn1cnc(c1Cl)[N+](=O)[O-]
IUPAC Name 5-chloro-1-methyl-4-nitroimidazole
Molecular Weight (g/mol) 161.55
Quantity 4 x 25 mg
Formula Weight 160.9992 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.